Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | deoxyhypusine hydroxylase/monooxygenase | DOHH |
HLRC1, hDOHH |
-19.5kb | Sense | Chr. 19 - 3490821 - 3500940 on NCBI Build GRCh38 |
| Downstream | major facilitator superfamily domain containing 12 | MFSD12 |
C19orf28, PP3501 |
9.6kb | Sense | Chr. 19 - 3538270 - 3557592 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Genomic Map