Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | ubiquitin specific peptidase 37 | USP37 | Sense | Chr. 2 - 218450251 - 218568393 on NCBI Build GRCh38 | ||
| Downstream | CCR4-NOT transcription complex subunit 9 | CNOT9 |
CAF40, CT129, RCD-1, RCD1, RQCD1 |
29.2kb | Anti-sense | Chr. 2 - 218568579 - 218596435 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Genomic Map