Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | reprimo like | RPRML | -155.6kb | Sense | Chr. 17 - 46978156 - 46979248 on NCBI Build GRCh38 | |
| Downstream | ribosomal protein S2 pseudogene 47 | RPS2P47 | RPS2_25_1553 | 23.3kb | Sense | Chr. 17 - 47159154 - 47159998 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Genomic Map