Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | transmembrane phosphoinositide 3-phosphatase and tensin homolog 2 pseudogene 5 | TPTE2P5 | Sense | Chr. 13 - 40796985 - 40921750 on NCBI Build GRCh38 | ||
| Downstream | E74 like ETS transcription factor 1 | ELF1 | 17.0kb | Sense | Chr. 13 - 40931919 - 41061414 on NCBI Build GRCh38 |
More Information
Set Membership: |
|

Genomic Map