Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream |
5',3'-nucleotidase, mitochondrial |
NT5M |
dNT-2, dNT2, mdN |
-137.4kb | Sense | Chr. 17 - 17303366 - 17347663 on NCBI Build GRCh38 |
| Downstream | ras related dexamethasone induced 1 | RASD1 |
AGS1, DEXRAS1, MGC:26290 |
6.1kb | Anti-sense | Chr. 17 - 17494437 - 17496395 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Genomic Map