Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | ubiquitin conjugating enzyme E2 J1 | UBE2J1 |
CGI-76, HSPC153, HSPC205, HSU93243, NCUBE-1, NCUBE1, UBC6, UBC6E, Ubc6p |
-7.2kb | Sense | Chr. 6 - 89326625 - 89352917 on NCBI Build GRCh38 |
| Downstream | Ras related GTP binding D | RRAGD |
RAGD, bA11D8.2.1 |
1.3kb | Sense | Chr. 6 - 89363509 - 89412470 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Genomic Map