Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | sulfatase 2 | SULF2 | HSULF-2 | Anti-sense | Chr. 20 - 47654584 - 47786629 on NCBI Build GRCh38 | |
| Downstream | uncharacterized LOC105372637 | LOC105372637 | 264.5kb | Sense | Chr. 20 - 47950810 - 47961246 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Genomic Map