Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | microRNA 3175 | MIR3175 | mir-3175 | -1.3kb | Sense | Chr. 15 - 92904399 - 92904475 on NCBI Build GRCh38 |
| Downstream | repulsive guidance molecule family member a | RGMA | RGM | 134.3kb | Anti-sense | Chr. 15 - 93043406 - 93089214 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Genomic Map