Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | ENO1 antisense RNA 1 | ENO1-AS1 | -81.6kb | Sense | Chr. 1 - 8878835 - 8879885 on NCBI Build GRCh38 | |
| Downstream | solute carrier family 2 member 5 | SLC2A5 |
GLUT-5, GLUT5 |
72.8kb | Anti-sense | Chr. 1 - 9036941 - 9093407 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Genomic Map