Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | uncharacterized LOC105374952 | LOC105374952 | -170.9kb | Anti-sense | Chr. 6 - 17706130 - 17711027 on NCBI Build GRCh38 | |
| Downstream | NHL repeat containing E3 ubiquitin protein ligase 1 | NHLRC1 |
EPM2A, EPM2B, MALIN, bA204B7.2 |
238.1kb | Sense | Chr. 6 - 18120487 - 18122620 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Genomic Map