Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | yippee like 1 | YPEL1 | FKSG3 | -33.9kb | Sense | Chr. 22 - 21697536 - 21735834 on NCBI Build GRCh38 |
| Downstream |
protein phosphatase, Mg2+/Mn2+ dependent 1F |
PPM1F |
CAMKP, CaMKPase, FEM-2, POPX2, hFEM-2 |
149.1kb | Sense | Chr. 22 - 21919419 - 21952878 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Genomic Map