Assay Details
| Gene Symbol: LOC105374896 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK130981.1 | - | 1 | 140 | 126 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | uncharacterized LOC105374895 | LOC105374895 | -107.6kb | Anti-sense | Chr. 6 - 4487714 - 4495771 on NCBI Build GRCh38 | |
| Downstream | uncharacterized LOC497048 | KU-MEL-3 | 8.1kb | Anti-sense | Chr. 6 - 4610565 - 4611919 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.
Set Membership: |
|

LOC105374896
Genomic Map