Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | uncharacterized LOC729083 | LOC729083 | -21.6kb | Anti-sense | Chr. 3 - 42770612 - 42773635 on NCBI Build GRCh38 | |
| Downstream | atypical chemokine receptor 2 | ACKR2 |
CCBP2, CCR10, CCR9, CMKBR9, D6, hD6 |
12.6kb | Anti-sense | Chr. 3 - 42809472 - 42867283 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Genomic Map