Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | long intergenic non-protein coding RNA 1426 | LINC01426 | lincRNA-uc002yug.2 | -12.5kb | Sense | Chr. 21 - 34745825 - 34784886 on NCBI Build GRCh38 |
| Downstream | RUNX1 intronic transcript 1 | RUNX1-IT1 | C21orf96 | 239.8kb | Anti-sense | Chr. 21 - 35037925 - 35039426 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.
Set Membership: |
|

Genomic Map