Assay Details
| Gene Symbol: COX10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | BC017752.1 | - | 3 | 1149 | 113 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | COX10 antisense RNA 1 | COX10-AS1 |
COX10-AS, COX10AS |
-147.2kb | Anti-sense | Chr. 17 - 14029292 - 14069458 on NCBI Build GRCh38 |
| Downstream | CMT1A duplicated region transcript 15 | CDRT15 | 17.9kb | Anti-sense | Chr. 17 - 14235856 - 14236833 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.
Set Membership: |
|

COX10
Genomic Map