Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | leucine rich repeat containing 8 family member D | LRRC8D | LRRC5 | -5.5kb | Sense | Chr. 1 - 89821014 - 89936432 on NCBI Build GRCh38 |
| Downstream | gem nuclear organelle associated protein 8 pseudogene 4 | GEMIN8P4 | 50.5kb | Anti-sense | Chr. 1 - 89993265 - 89994966 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Genomic Map