Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | upstream transcription factor family member 3 | USF3 | KIAA2018 | -16.7kb | Sense | Chr. 3 - 113648385 - 113696657 on NCBI Build GRCh38 |
| Downstream |
N(alpha)-acetyltransferase 50, NatE catalytic subunit |
NAA50 |
MAK3, NAT13, NAT13P, NAT5, NAT5P, SAN |
2.0kb | Sense | Chr. 3 - 113716460 - 113746299 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Genomic Map