Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | matrix metallopeptidase 16 | MMP16 |
C8orf57, MMP-X2, MT-MMP2, MT-MMP3, MT3-MMP |
-408.4kb | Anti-sense | Chr. 8 - 88037232 - 88327488 on NCBI Build GRCh38 |
| Downstream | uncharacterized LOC101929709 | LOC101929709 | 980.3kb | Anti-sense | Chr. 8 - 89717399 - 89757711 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Genomic Map