Assay Details
| Gene Symbol: LOC101928718 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | BC035160.2 | - | 1 | 1449 | 100 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | FAM212B antisense RNA 1 | FAM212B-AS1 | -6.0kb | Anti-sense | Chr. 1 - 111739841 - 111747798 on NCBI Build GRCh38 | |
| Downstream | DEAD-box helicase 20 | DDX20 |
DP103, GEMIN3 |
31.0 | Anti-sense | Chr. 1 - 111755568 - 111767577 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0100015287.hg.1 | ENST00000412270.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015287.hg.1 | ENST00000412270.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015287.hg.1 | ENST00000412270.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015287.hg.1 | ENST00000412270.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015287.hg.1 | ENST00000412270.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015287.hg.1 | ENST00000412270.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015287.hg.1 | ENST00000527660.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015287.hg.1 | ENST00000527660.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015287.hg.1 | ENST00000625113.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015287.hg.1 | ENST00000625113.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015287.hg.1 | ENST00000625113.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015287.hg.1 | NR_125963.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015287.hg.1 | NR_125963.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015287.hg.1 | NR_125963.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015287.hg.1 | NR_125963.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015287.hg.1 | NR_125963.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015287.hg.1 | NR_125963.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009428.hg.1 | ENST00000369702.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009428.hg.1 | ENST00000369702.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009428.hg.1 | ENST00000369702.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009428.hg.1 | ENST00000369702.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009428.hg.1 | ENST00000369702.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009428.hg.1 | ENST00000369702.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009428.hg.1 | ENST00000369702.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009428.hg.1 | ENST00000369702.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009428.hg.1 | ENST00000369702.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009428.hg.1 | ENST00000369702.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009428.hg.1 | ENST00000369702.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009428.hg.1 | ENST00000369702.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009428.hg.1 | ENST00000369702.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009428.hg.1 | ENST00000369702.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009428.hg.1 | ENST00000369702.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009428.hg.1 | ENST00000369702.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009428.hg.1 | ENST00000369702.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009428.hg.1 | ENST00000369702.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009428.hg.1 | ENST00000369702.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009428.hg.1 | ENST00000369702.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009428.hg.1 | ENST00000369702.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009428.hg.1 | ENST00000369702.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009428.hg.1 | ENST00000475700.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009428.hg.1 | ENST00000475700.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009428.hg.1 | ENST00000475700.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009428.hg.1 | ENST00000475700.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009428.hg.1 | ENST00000475700.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009428.hg.1 | ENST00000475700.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009428.hg.1 | ENST00000475700.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009428.hg.1 | ENST00000475700.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009428.hg.1 | ENST00000475700.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009428.hg.1 | ENST00000475700.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009428.hg.1 | ENST00000475700.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009428.hg.1 | ENST00000475700.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009428.hg.1 | ENST00000475700.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009428.hg.1 | ENST00000475700.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009428.hg.1 | ENST00000475700.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009428.hg.1 | ENST00000475700.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009428.hg.1 | ENST00000475700.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009428.hg.1 | ENST00000475700.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009428.hg.1 | ENST00000475700.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009428.hg.1 | ENST00000475700.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009428.hg.1 | ENST00000475700.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009428.hg.1 | ENST00000524894.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009428.hg.1 | ENST00000524894.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009428.hg.1 | ENST00000524894.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009428.hg.1 | ENST00000524894.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009428.hg.1 | ENST00000524894.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009428.hg.1 | ENST00000533164.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009428.hg.1 | ENST00000533164.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009428.hg.1 | ENST00000533164.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009428.hg.1 | ENST00000533164.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009428.hg.1 | ENST00000533164.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009428.hg.1 | ENST00000533164.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009428.hg.1 | ENST00000533164.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009428.hg.1 | ENST00000533164.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009428.hg.1 | ENST00000533164.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009428.hg.1 | ENST00000533164.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009428.hg.1 | ENST00000533164.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009428.hg.1 | ENST00000533164.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009428.hg.1 | ENST00000533164.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009428.hg.1 | ENST00000533164.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009428.hg.1 | ENST00000534200.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009428.hg.1 | ENST00000534200.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009428.hg.1 | ENST00000534200.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009428.hg.1 | ENST00000534200.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009428.hg.1 | ENST00000534200.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009428.hg.1 | NM_007204.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009428.hg.1 | NM_007204.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009428.hg.1 | NM_007204.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009428.hg.1 | NM_007204.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009428.hg.1 | NM_007204.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009428.hg.1 | NM_007204.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009428.hg.1 | NM_007204.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009428.hg.1 | NM_007204.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009428.hg.1 | NM_007204.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009428.hg.1 | NM_007204.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009428.hg.1 | NM_007204.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009428.hg.1 | NM_007204.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009428.hg.1 | NM_007204.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009428.hg.1 | NM_007204.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009428.hg.1 | NM_007204.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009428.hg.1 | NM_007204.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009428.hg.1 | NM_007204.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009428.hg.1 | NM_007204.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009428.hg.1 | NM_007204.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009428.hg.1 | ENST00000369702.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009428.hg.1 | ENST00000369702.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009428.hg.1 | ENST00000369702.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009428.hg.1 | ENST00000369702.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009428.hg.1 | ENST00000369702.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009428.hg.1 | ENST00000369702.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009428.hg.1 | ENST00000369702.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009428.hg.1 | ENST00000369702.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009428.hg.1 | ENST00000369702.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009428.hg.1 | ENST00000369702.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009428.hg.1 | ENST00000369702.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009428.hg.1 | ENST00000369702.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009428.hg.1 | ENST00000369702.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009428.hg.1 | ENST00000369702.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009428.hg.1 | ENST00000369702.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009428.hg.1 | ENST00000369702.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009428.hg.1 | ENST00000369702.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009428.hg.1 | ENST00000369702.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009428.hg.1 | ENST00000369702.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009428.hg.1 | ENST00000369702.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009428.hg.1 | ENST00000369702.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009428.hg.1 | ENST00000369702.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009428.hg.1 | ENST00000475700.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009428.hg.1 | ENST00000475700.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009428.hg.1 | ENST00000475700.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009428.hg.1 | ENST00000475700.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009428.hg.1 | ENST00000475700.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009428.hg.1 | ENST00000475700.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009428.hg.1 | ENST00000475700.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009428.hg.1 | ENST00000475700.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009428.hg.1 | ENST00000475700.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009428.hg.1 | ENST00000475700.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009428.hg.1 | ENST00000475700.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009428.hg.1 | ENST00000475700.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009428.hg.1 | ENST00000475700.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009428.hg.1 | ENST00000475700.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009428.hg.1 | ENST00000475700.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009428.hg.1 | ENST00000475700.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009428.hg.1 | ENST00000475700.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009428.hg.1 | ENST00000475700.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009428.hg.1 | ENST00000475700.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009428.hg.1 | ENST00000475700.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009428.hg.1 | ENST00000475700.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009428.hg.1 | ENST00000524894.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009428.hg.1 | ENST00000524894.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009428.hg.1 | ENST00000524894.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009428.hg.1 | ENST00000524894.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009428.hg.1 | ENST00000524894.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009428.hg.1 | ENST00000533164.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009428.hg.1 | ENST00000533164.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009428.hg.1 | ENST00000533164.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009428.hg.1 | ENST00000533164.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009428.hg.1 | ENST00000533164.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009428.hg.1 | ENST00000533164.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009428.hg.1 | ENST00000533164.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009428.hg.1 | ENST00000533164.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009428.hg.1 | ENST00000533164.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009428.hg.1 | ENST00000533164.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009428.hg.1 | ENST00000533164.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009428.hg.1 | ENST00000533164.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009428.hg.1 | ENST00000533164.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009428.hg.1 | ENST00000533164.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009428.hg.1 | ENST00000534200.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009428.hg.1 | ENST00000534200.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009428.hg.1 | ENST00000534200.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009428.hg.1 | ENST00000534200.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009428.hg.1 | ENST00000534200.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009428.hg.1 | NM_007204.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009428.hg.1 | NM_007204.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009428.hg.1 | NM_007204.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009428.hg.1 | NM_007204.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009428.hg.1 | NM_007204.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009428.hg.1 | NM_007204.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009428.hg.1 | NM_007204.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009428.hg.1 | NM_007204.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009428.hg.1 | NM_007204.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009428.hg.1 | NM_007204.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009428.hg.1 | NM_007204.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009428.hg.1 | NM_007204.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009428.hg.1 | NM_007204.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009428.hg.1 | NM_007204.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009428.hg.1 | NM_007204.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009428.hg.1 | NM_007204.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009428.hg.1 | NM_007204.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009428.hg.1 | NM_007204.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009428.hg.1 | NM_007204.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015287.hg.1 | ENST00000412270.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015287.hg.1 | ENST00000412270.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015287.hg.1 | ENST00000412270.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015287.hg.1 | ENST00000412270.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015287.hg.1 | ENST00000412270.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015287.hg.1 | ENST00000412270.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015287.hg.1 | ENST00000527660.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015287.hg.1 | ENST00000527660.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015287.hg.1 | ENST00000625113.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015287.hg.1 | ENST00000625113.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015287.hg.1 | ENST00000625113.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015287.hg.1 | NR_125963.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015287.hg.1 | NR_125963.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015287.hg.1 | NR_125963.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015287.hg.1 | NR_125963.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015287.hg.1 | NR_125963.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015287.hg.1 | NR_125963.1 | Refseq |

LOC101928718
Genomic Map