Assay Details
| Gene Symbol: LOC105375240 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | BC036546.1 | - | 1 | 351 | 101 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | POU class 6 homeobox 2 | POU6F2 |
RPF-1, WT5, WTSL |
-94.4kb | Anti-sense | Chr. 7 - 38977878 - 39468601 on NCBI Build GRCh38 |
| Downstream | Yae1 domain containing 1 | YAE1D1 |
C7orf36, GK003 |
23.0 | Anti-sense | Chr. 7 - 39566376 - 39612089 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.
Set Membership: |
|

LOC105375240
Genomic Map