Assay Details
| Gene Symbol: KIN | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_012311.3 | NP_036443.1 | 13 | 6203 | 118 | |
| NR_045609.1 | 14 | 3778 | 118 | |||
| GenBank mRNA | AA768850.1 | - | 1 | 118 | ||
| BC037913.2 | - | 2 | 2352 | 118 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | inter-alpha-trypsin inhibitor heavy chain 2 | ITIH2 |
H2P, SHAP |
-1.4kb | Anti-sense | Chr. 10 - 7703273 - 7749520 on NCBI Build GRCh38 |
| Downstream |
ATP synthase, H+ transporting, mitochondrial F1 complex, gamma polypeptide 1 |
ATP5C1 |
ATP5C, ATP5CL1 |
36.3kb | Anti-sense | Chr. 10 - 7788104 - 7807801 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

KIN
Genomic Map