Assay Details
| Gene Symbol: LOC101929319 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_110248.1 | 3 | 1247 | 101 | ||
| GenBank mRNA | BC039384.1 | - | 3 | 1247 | 101 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | N-myc and STAT interactor | NMI | -47.9kb | Sense | Chr. 2 - 151270465 - 151289916 on NCBI Build GRCh38 | |
| Downstream | TNF alpha induced protein 6 | TNFAIP6 |
TSG-6, TSG6 |
18.2kb | Anti-sense | Chr. 2 - 151357592 - 151380048 on NCBI Build GRCh38 |
More Information
Updated Mapping Information
Set Membership: |
|

LOC101929319
Genomic Map