Assay Details
| Gene Symbol: LOC105372394 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | BC042546.1 | - | 2 | 3953 | 99 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | zinc finger protein 573 | ZNF573 | -28.2kb | Sense | Chr. 19 - 37738274 - 37779626 on NCBI Build GRCh38 | |
| Downstream | uncharacterized LOC644554 | LOC644554 | 8.5kb | Anti-sense | Chr. 19 - 37817411 - 37826638 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.
Set Membership: |
|

LOC105372394
Genomic Map