Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | UGDH antisense RNA 1 | UGDH-AS1 | -2.7kb | Anti-sense | Chr. 4 - 39527839 - 39594707 on NCBI Build GRCh38 | |
| Downstream | uncharacterized LOC105374419 | LOC105374419 | 38.7kb | Anti-sense | Chr. 4 - 39639095 - 39666653 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Genomic Map