Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | vesicle amine transport 1-like | VAT1L | -34.6kb | Anti-sense | Chr. 16 - 77788564 - 77980107 on NCBI Build GRCh38 | |
| Downstream | C-type lectin domain family 3 member A | CLEC3A | CLECSF1 | 7.4kb | Anti-sense | Chr. 16 - 78022546 - 78032106 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Genomic Map