Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | SRA stem-loop interacting RNA binding protein | SLIRP |
C14orf156, DC50, PD04872 |
-42.2kb | Anti-sense | Chr. 14 - 77708071 - 77717598 on NCBI Build GRCh38 |
| Downstream | aarF domain containing kinase 1 | ADCK1 | 38.9kb | Anti-sense | Chr. 14 - 77800058 - 77933954 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Genomic Map