Assay Details
| Gene Symbol: TTC3-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | BC066351.1 | - | 2 | 847 | 99 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | phosphatidylinositol glycan anchor biosynthesis class P | PIGP |
DCRC, DCRC-S, DSCR5, DSRC, PIG-P |
-114.5kb | Sense | Chr. 21 - 37065364 - 37073158 on NCBI Build GRCh38 |
| Downstream | Down syndrome critical region 9 (non-protein coding) | DSCR9 | NCRNA00038 | 19.9kb | Anti-sense | Chr. 21 - 37208503 - 37221736 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

TTC3-AS1
Genomic Map