Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | transmembrane protein 14B | TMEM14B | -33.1kb | Sense | Chr. 6 - 10747759 - 10759774 on NCBI Build GRCh38 | |
| Downstream | glial cells missing homolog 2 | GCM2 |
GCMB, hGCMb |
77.0kb | Anti-sense | Chr. 6 - 10873223 - 10881865 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Genomic Map