Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | SLC25A21 antisense RNA 1 | SLC25A21-AS1 | -234.6kb | Anti-sense | Chr. 14 - 37172026 - 37173794 on NCBI Build GRCh38 | |
| Downstream | forkhead box A1 | FOXA1 |
HNF3A, TCF3A |
176.7kb | Sense | Chr. 14 - 37589552 - 37595120 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Genomic Map