Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | uncharacterized LOC105370829 | LOC105370829 | -79.8kb | Anti-sense | Chr. 15 - 55056747 - 55092173 on NCBI Build GRCh38 | |
| Downstream | ribosomal L24 domain containing 1 | RSL24D1 |
C15orf15, HRP-L30-iso, L30, RLP24, RPL24, RPL24L, TVAS3 |
4.6kb | Sense | Chr. 15 - 55181314 - 55197033 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Genomic Map