Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | microRNA 6085 | MIR6085 | hsa-mir-6085 | -139.2kb | Sense | Chr. 15 - 62343029 - 62343138 on NCBI Build GRCh38 |
| Downstream | uncharacterized protein MGC15885 | MGC15885 | 148.2kb | Anti-sense | Chr. 15 - 62637172 - 62645181 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Genomic Map