Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | microRNA 3684 | MIR3684 | Sense | Chr. 4 - 98997387 - 98997460 on NCBI Build GRCh38 | ||
| Downstream |
alcohol dehydrogenase 5 (class III), chi polypeptide |
ADH5 |
ADH-3, ADHX, FALDH, FDH, GSH-FDH, GSNOR, HEL-S-60p |
72.8kb | Anti-sense | Chr. 4 - 99070978 - 99088788 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Genomic Map