Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | family with sequence similarity 69 member A | FAM69A | Sense | Chr. 1 - 92832729 - 92961522 on NCBI Build GRCh38 | ||
| Downstream | metal response element binding transcription factor 2 | MTF2 |
M96, PCL2, TDRD19A, dJ976O13.2 |
145.1kb | Anti-sense | Chr. 1 - 93079219 - 93139081 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Genomic Map