Assay Details
| Gene Symbol: SERPINE1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017012260.1 | XP_016867749.1 | 7 | 1317 | 93 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | tripartite motif containing 56 | TRIM56 | RNF109 | -44.0kb | Sense | Chr. 7 - 101085424 - 101097967 on NCBI Build GRCh38 |
| Downstream | adaptor related protein complex 1 sigma 1 subunit | AP1S1 |
AP19, CLAPS1, EKV3, MEDNIK, SIGMA1A |
18.3kb | Sense | Chr. 7 - 101154405 - 101161276 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

SERPINE1
Genomic Map