Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | proline-serine-threonine phosphatase interacting protein 2 | PSTPIP2 | MAYP | -11.9kb | Sense | Chr. 18 - 45983536 - 46072327 on NCBI Build GRCh38 |
| Downstream | HAUS augmin like complex subunit 1 | HAUS1 |
CCDC5, HEI-C, HEIC, HsT1461 |
15.6kb | Anti-sense | Chr. 18 - 46104332 - 46128333 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Genomic Map