Assay Details
| Gene Symbol: CRIM1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | CR627451.1 | - | 1 | 1551 | 83 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | uncharacterized LOC100288911 | LOC100288911 | -185.0kb | Anti-sense | Chr. 2 - 36354749 - 36355570 on NCBI Build GRCh38 | |
| Downstream | fasciculation and elongation protein zeta 2 | FEZ2 | HUM3CL | 9.6kb | Anti-sense | Chr. 2 - 36552239 - 36598190 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

CRIM1
Genomic Map