Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | ankyrin repeat and sterile alpha motif domain containing 1B | ANKS1B |
AIDA, AIDA-1, ANKS2, EB-1, EB1, cajalin-2 |
-107.8kb | Anti-sense | Chr. 12 - 98729904 - 99984727 on NCBI Build GRCh38 |
| Downstream | golgin A2 pseudogene 5 | GOLGA2P5 |
GOLGA2B, GOLGA2L1 |
60.4kb | Anti-sense | Chr. 12 - 100156391 - 100173343 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Genomic Map