Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | zinc finger protein 292 | ZNF292 |
Nbla00365, ZFP292, ZN-16, Zn-15, bA393I2.3 |
Sense | Chr. 6 - 87155551 - 87264172 on NCBI Build GRCh38 | |
| Downstream | gap junction protein beta 7 | GJB7 |
CX25, bA136M9.1, connexin25 |
119.9kb | Anti-sense | Chr. 6 - 87282978 - 87329278 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Genomic Map