Assay Details
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Transcript Location | Amplicon Length | |
|---|---|---|---|---|---|---|---|
| GenBankmRNA | DQ655978.1 | - | 2 | 849 | Chr. 16 - 47236 - 52142 [+] on Build GRCh38 | 81 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | WASH and IL9R antisense RNA 2 | WASIR2 | NCRNA00286A | -26.4kb | Sense | Chr. 16 - 22916 - 25114 on NCBI Build GRCh38 |
| Downstream | small nuclear ribonucleoprotein U11/U12 subunit 25 | SNRNP25 | C16orf33 | 1.7kb | Sense | Chr. 16 - 53829 - 57669 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Genomic Map