Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | TATA-box binding protein associated factor 7 like | TAF7L |
CT40, TAF2Q |
-55.5kb | Anti-sense | Chr. X - 101268253 - 101293071 on NCBI Build GRCh38 |
| Downstream | Bruton tyrosine kinase | BTK |
AGMX1, AT, ATK, BPK, IMD1, PSCTK1, XLA |
251.0 | Anti-sense | Chr. X - 101349447 - 101390796 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Genomic Map