Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | PHD finger protein 6 | PHF6 |
BFLS, BORJ, CENP-31 |
-64.8kb | Sense | Chr. X - 134373312 - 134428792 on NCBI Build GRCh38 |
| Downstream | microRNA 450b | MIR450B |
MIRN450B, mir-450b |
46.4kb | Anti-sense | Chr. X - 134540185 - 134540262 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Genomic Map