Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | glycogenin 2 | GYG2 |
GN-2, GN2 |
Sense | Chr. X - 2828712 - 2882820 on NCBI Build GRCh38 | |
| Downstream | arylsulfatase D | ARSD | ASD | 22.4kb | Anti-sense | Chr. X - 2903970 - 2929375 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Genomic Map