Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | BCL2 like 13 | BCL2L13 |
BCL-RAMBO, Bcl2-L-13, MIL1 |
Sense | Chr. 22 - 17628855 - 17730855 on NCBI Build GRCh38 | |
| Downstream | BH3 interacting domain death agonist | BID | FP497 | 46.8kb | Anti-sense | Chr. 22 - 17734140 - 17774665 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Genomic Map