Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | NADH:ubiquinone oxidoreductase complex assembly factor 3 | NDUFAF3 |
2P1, C3orf60, E3-3 |
-178.0 | Anti-sense | Chr. 3 - 49020475 - 49023495 on NCBI Build GRCh38 |
| Downstream | glutamine rich 1 | QRICH1 | 5.3kb | Sense | Chr. 3 - 49029707 - 49094373 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Genomic Map