Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream |
RFX family member 8, lacking RFX DNA binding domain |
RFX8 | -257.2kb | Anti-sense | Chr. 2 - 101397344 - 101475179 on NCBI Build GRCh38 | |
| Downstream | long intergenic non-protein coding RNA 1127 | LINC01127 | 250.8kb | Sense | Chr. 2 - 101983465 - 101985545 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Genomic Map