Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | H2A histone family member Y2 | H2AFY2 | macroH2A2 | Sense | Chr. 10 - 70052601 - 70112284 on NCBI Build GRCh38 | |
| Downstream |
apoptosis inducing factor, mitochondria associated 2 |
AIFM2 |
AMID, PRG3 |
26.0kb | Anti-sense | Chr. 10 - 70112267 - 70132934 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Genomic Map