Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | complement component 3 precursor pseudogene | C3P1 | CPLP | -15.3kb | Sense | Chr. 19 - 10041356 - 10074138 on NCBI Build GRCh38 |
| Downstream | angiopoietin like 6 | ANGPTL6 |
AGF, ARP5 |
2.0kb | Anti-sense | Chr. 19 - 10092337 - 10106194 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Genomic Map