Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | phosphatase and actin regulator 4 | PHACTR4 | PPP1R124 | Sense | Chr. 1 - 28369574 - 28500369 on NCBI Build GRCh38 | |
| Downstream | regulator of chromosome condensation 1 | RCC1 |
CHC1, RCC1-I, SNHG3-RCC1 |
35.9kb | Sense | Chr. 1 - 28505943 - 28539196 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Genomic Map