Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | uncharacterized LOC101928471 | LOC101928471 | -9.9kb | Sense | Chr. 12 - 24223271 - 24237965 on NCBI Build GRCh38 | |
| Downstream | long intergenic non-protein coding RNA 477 | LINC00477 |
C12orf67, FAM191B |
318.7kb | Anti-sense | Chr. 12 - 24566964 - 24584168 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Genomic Map