Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | SRD5A3 antisense RNA 1 | SRD5A3-AS1 | -12.8kb | Anti-sense | Chr. 4 - 55366601 - 55385580 on NCBI Build GRCh38 | |
| Downstream | clock circadian regulator | CLOCK |
KAT13D, bHLHe8 |
29.0kb | Anti-sense | Chr. 4 - 55427901 - 55547138 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Genomic Map